2-({2-[(2,6-Dichloropyridin-3-yl)amino]-2-oxoethyl}thio)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-({[(Carboxymethyl)thio]acetyl}amino)-2,6-dichloropyridine, 5-{[(Carboxymethyl)sulphanyl]acetamido}-2,6-dichloropyridine
OR27325-250MG
APOSOR27325-250MGEA
46.4
EUR
APOSOR27325-250MG
2-({2-[(2,6-Dichloropyridin-3-yl)amino]-2-oxoethyl}thio)acetic acid
2-({2-[(2,6-Dichloropyridin-3-yl)amino]-2-oxoethyl}thio)acetic acid
Formel:
C₉H₈Cl₂N₂O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662316 CAS-Nummer: 287197-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...