2-(3,5-Dichlorophenyl)propene 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR2733-1G
APOSOR2733-1GEA
0
EUR
APOSOR2733-1G
2-(3,5-Dichlorophenyl)propene 99%
2-(3,5-Dichlorophenyl)propene
Formel:
C₉H₈Cl₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035987 CAS-Nummer: 68575-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...