2-Amino-2-thioxoethyl pivalate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-2-thioxoethyl 2,2-dimethylpropanoate

OR27343-1G
APOSOR27343-1GEA 74.1 EUR
APOSOR27343-1G
2-Amino-2-thioxoethyl pivalate
2-Amino-2-thioxoethyl pivalate
Formel: C₇H₁₃NO₂S
Molecular Weight: 175.25 g/mol
Schmelzpunkt: 84…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00204238
CAS-Nummer: 175204-79-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR