1H-Pyrrole-2-carbohydrazide

Lieferant: Apollo Scientific

OR27344-1G
APOSOR27344-1GEA 87 EUR
APOSOR27344-1G
1H-Pyrrole-2-carbohydrazide
1H-Pyrrole-2-carbohydrazide
Formel: C₅H₇N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00141885
CAS-Nummer: 50269-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR