5-(Methoxycarbonyl)-2-pyridine carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Methoxycarbonyl)picolinic acid

OR27346-1G
APOSOR27346-1GEA 50.9 EUR
APOSOR27346-1G
5-(Methoxycarbonyl)-2-pyridine carboxylic acid
5-(Methoxycarbonyl)-2-pyridine carboxylic acid
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00179343
CAS-Nummer: 17874-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR