3-Hydroxy-7-methyl-1,2-benzisoxazole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Methylbenzo[d]isoxazol-3-ol, 7-Methyl-1,2-benzisoxazol-3-ol, 7-Methylbenzo[d]isoxazol-3(2H)-one

OR27361-50MG
APOSOR27361-50MGEA 21.9 EUR
APOSOR27361-50MG
3-Hydroxy-7-methyl-1,2-benzisoxazole 97%
3-Hydroxy-7-methyl-1,2-benzisoxazole
Formel: C₈H₇NO₂
Molecular Weight: 149.147 g/mol
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00104669
CAS-Nummer: 36238-83-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR