2-Chloro-6-methoxyisonicotinoyl chloride

Lieferant: Apollo Scientific
Gefahr

OR27364-1G OR27364-250MG
APOSOR27364-1GEA 60.9 EUR
APOSOR27364-1G APOSOR27364-250MG
2-Chloro-6-methoxyisonicotinoyl chloride
2-Chloro-6-methoxyisonicotinoyl chloride
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206.03 g/mol
Siedepunkt: 63…64 °C/0.01mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00125355
CAS-Nummer: 116853-97-5
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR