6-Methoxynicotinonitril 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Methoxypyridine-5-carbonitrile

OR2738-25G
APOSOR2738-25GEA 198 EUR
APOSOR2738-25G
6-Methoxynicotinonitril 97%
6-Methoxynicotinonitril
Formel: C₇H₆N₂O
Molecular Weight: 134.1400 g/mol
Schmelzpunkt: 93…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084981
CAS-Nummer: 15871-85-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR