Pyrazole-3-carboxaldehyde dimethyl acetal

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Dimethoxymethyl)-1H-pyrazole

OR27386-1G
APOSOR27386-1GEA 45.1 EUR
APOSOR27386-1G
Pyrazole-3-carboxaldehyde dimethyl acetal
Pyrazole-3-carboxaldehyde dimethyl acetal
Formel: C₆H₁₀N₂O₂
Molecular Weight: 142,1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085025
CAS-Nummer: 111573-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR