4-Methylnicotinamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methyl-3-pyridinecarboxamide

OR27413-1G
APOSOR27413-1GEA 236 EUR
APOSOR27413-1G
4-Methylnicotinamid
4-Methylnicotinamid
Formel: C₇H₈N₂O
Molecular Weight: 136,1512 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234322
CAS-Nummer: 7250-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR