2-(2-Isocyanatoethyl)thiophene

Lieferant: Apollo Scientific

Synonyme: 2-(Thien-2-yl)ethyl isocyanate

OR27429-1G
APOSOR27429-1GEA 0 EUR
APOSOR27429-1G
2-(2-Isocyanatoethyl)thiophene
2-(2-Isocyanatoethyl)thiophene

May form small amount of sediment

Formel: C₇H₇NOS
Molecular Weight: 153.202 g/mol
Siedepunkt: 102 °C (15 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052972
CAS-Nummer: 58749-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR