2,6-Dibrom-p-toluidin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dibrom-4-methylanilin, 4-Amino-3,5-dibromtoluol

OR27443-5G
APOSOR27443-5GEA 21.9 EUR
APOSOR27443-5G
2,6-Dibrom-p-toluidin
2,6-Dibrom-p-toluidin
Formel: C₇H₇Br₂N
Molecular Weight: 264,95 g/mol
Schmelzpunkt: 72…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007641
CAS-Nummer: 6968-24-7
EINECS: 230-182-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR