Ethyl quinoxaline-6-carboxylate

Lieferant: Apollo Scientific
Achtung

OR2746-250MG
APOSOR2746-250MGEA 21.9 EUR
APOSOR2746-250MG
Ethyl quinoxaline-6-carboxylate
Ethyl quinoxaline-6-carboxylate
Formel: C₁₁H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09998746
CAS-Nummer: 6924-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR