6-Nitropiperonal 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Methylendioxy-6-nitrobenzaldehyd, 6-Nitro-1,3-benzodioxol-5-carbaldehyd

OR27479-1G
APOSOR27479-1GEA 31.9 EUR
APOSOR27479-1G
6-Nitropiperonal 98%
6-Nitropiperonal
Formel: C₈H₅NO₅
Molecular Weight: 195,13 g/mol
Schmelzpunkt: 93…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005829
CAS-Nummer: 712-97-0
EINECS: 211-926-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR