6-Bromo-8-nitroimidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR2749-1G
APOSOR2749-1GEA
145
EUR
APOSOR2749-1G
6-Bromo-8-nitroimidazo[1,2-a]pyridine
6-Bromo-8-nitroimidazo[1,2-a]pyridine
Formel:
C₇H₄BrN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836167 CAS-Nummer: 1031927-16-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...