Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR2750-1G
APOSOR2750-1GEA
37.7
EUR
APOSOR2750-1G
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Methyl-3-bromimidazo[1,2-a]pyridin-6-carboxylat
Formel:
C₉H₇BrN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021501 CAS-Nummer: 886361-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...