8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR2753-1G
APOSOR2753-1GEA 346 EUR
APOSOR2753-1G
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Formel: C₈H₆BrClN₄O
Molecular Weight: 289,53 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09951830
CAS-Nummer: 1031927-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR