2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
Lieferant: Apollo Scientific
Synonyme:
1,3-Di(tert-butyl)-2-hydroxy-5-methylcyclohexane, 2,6-Di-tert-butyl-4-methylcyclohexanol
OR27530-25G
APOSOR27530-25GEA
62.5
EUR
APOSOR27530-25G
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol
Formel:
C₁₅H₃₀O Molecular Weight: 226,398 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00278493 CAS-Nummer: 163119-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...