5-Iodindol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Jodindol, 5-Jod-1H-indol

OR27541-250MG
APOSOR27541-250MGEA 21.9 EUR
APOSOR27541-250MG
5-Iodindol 97%
5-Iodindol
Formel: C₈H₆IN
Molecular Weight: 243,05 g/mol
Schmelzpunkt: 101…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00220065
CAS-Nummer: 16066-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR