Ethyl 5-Nitrobenzofuran-2-Carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Nitrobenzofuran-2-carboxylic Acid Ethyl Ester, Ethyl 5-nitrobenzofuran-2-carboxylate

OR2755-1G
APOSOR2755-1GEA 21.9 EUR
APOSOR2755-1G
Ethyl 5-Nitrobenzofuran-2-Carboxylat
Ethyl-5-nitrobenzofuran-2-carboxylate
Formel: C₁₁H₉NO₅
Molecular Weight: 235,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00275272
CAS-Nummer: 69604-00-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR