trans-3-Ethoxycinnamic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (2E)-3-(3-Ethoxyphenyl)prop-2-enoic acid, (E)-3-(3-Ethoxyphenyl)acrylic acid

OR27554-1G
APOSOR27554-1GEA 136 EUR
APOSOR27554-1G
trans-3-Ethoxycinnamic acid
trans-3-Ethoxycinnamic acid
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Schmelzpunkt: 131…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016843
CAS-Nummer: 188545-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR