5-Aminobenzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-1H-benzimidazole
OR27567-1G
APOSOR27567-1GEA
21.9
EUR
APOSOR27567-1G
5-Aminobenzimidazole
5-Aminobenzimidazole
Formel:
C₇H₇N₃ Molecular Weight: 133.15 g/mol Schmelzpunkt: 163…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00465258 CAS-Nummer: 934-22-5 EINECS: 213-279-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...