2-Aminothiazolinone hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Aminothiazol-4(5H)-one hydrochloride
OR27571-1G
APOSOR27571-1GEA
21.9
EUR
APOSOR27571-1G
2-Aminothiazolinone hydrochloride
2-Aminothiazolinone hydrochloride
Formel:
C₃H₅ClN₂OS Molecular Weight: 152,6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00277945 CAS-Nummer: 2192-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...