Methyl-4-hydroxycyclohexanecarboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR27594-1G OR27594-5G
APOSOR27594-1GEA 0 EUR
APOSOR27594-1G APOSOR27594-5G
Methyl-4-hydroxycyclohexanecarboxylate 95%
Methyl-4-hydroxycyclohexanecarboxylate
Formel: C₈H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00729087
CAS-Nummer: 17449-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR