4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N,N-Dimethyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride

OR2765-500MG
APOSOR2765-500MGEA 36.4 EUR
APOSOR2765-500MG
4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride
4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride
Formel: C₁₅H₂₄BNO₂·ClH
Molecular Weight: 297,628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07771993
CAS-Nummer: 1073371-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR