1-Methyloxindol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methylindolin-2-one

OR27652-5G
APOSOR27652-5GEA 97.3 EUR
APOSOR27652-5G
1-Methyloxindol 98%
1-Methyloxindol
Formel: C₉H₉NO
Molecular Weight: 147,1739 g/mol
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00030253
CAS-Nummer: 61-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR