6-(Dimethylamino)pyridine-3-boronic acid hydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Borono-2-(dimethylamino)pyridine hydrate

OR2767-1G OR2767-500MG
APOSOR2767-1GEA 69.6 EUR
APOSOR2767-1G APOSOR2767-500MG
6-(Dimethylamino)pyridine-3-boronic acid hydrate
6-(Dimethylamino)pyridine-3-boronic acid hydrate
Formel: C₇H₁₁BN₂O₂·H₂O
Lagertemperatur: Kühlschrank
MDL: MFCD06656059
CAS-Nummer: 1256355-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR