2-(Dimethylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: Apollo Scientific

OR2768-1G OR2768-5G OR2768-25G
APOSOR2768-1GEA 0 EUR
APOSOR2768-1G APOSOR2768-5G APOSOR2768-25G
2-(Dimethylamino)-5-pyrimidinboronsäurepinakolester
2-(Dimethylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₂H₂₀BN₃O₂
Schmelzpunkt: 107…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07368238
CAS-Nummer: 1032759-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR