2-Hydroxy-1,3,5-benzenetricarbaldehyde 95+%
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxybenzene-1,3,5-tricarbaldehyde
OR27683-1G
APOSOR27683-1GEA
55.1
EUR
APOSOR27683-1G
2-Hydroxy-1,3,5-benzenetricarbaldehyde 95+%
2-Hydroxy-1,3,5-benzenetricarbaldehyde
Formel:
C₉H₆O₄ Molecular Weight: 178,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01314207 CAS-Nummer: 81502-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...