(4-Methoxybenzyl)triphenylphosphoniumbromid

Lieferant: Apollo Scientific
Achtung

OR27696-1G
APOSOR27696-1GEA 186 EUR
APOSOR27696-1G
(4-Methoxybenzyl)triphenylphosphoniumbromid
(4-Methoxybenzyl)triphenylphosphoniumbromid
Formel: C₂₆H₂₄BrOP
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1530-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR