2,6-Dichlor-3-nitrobenzaldehyd
Lieferant: Apollo Scientific
|
Gefahr
|
OR27698-1G
APOSOR27698-1GEA
21.9
EUR
APOSOR27698-1G
2,6-Dichlor-3-nitrobenzaldehyd
2,6-Dichloro-3-nitrobenzaldehyde
Formel:
C₇H₃Cl₂NO₃ Molecular Weight: 220,0096 g/mol Schmelzpunkt: 73…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090332 CAS-Nummer: 5866-97-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...