2,6-Dichloro-3-nitrobenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

OR27700-1G
APOSOR27700-1GEA 21.9 EUR
APOSOR27700-1G
2,6-Dichloro-3-nitrobenzyl alcohol 97%
2,6-Dichloro-3-nitrobenzyl alcohol
Formel: C₇H₅Cl₂NO₃
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01566901
CAS-Nummer: 160647-01-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR