2-Chloro-N-cyclohexyl-N-phenylacetamide

Lieferant: Apollo Scientific

OR27702-1G
APOSOR27702-1GEA 0 EUR
APOSOR27702-1G
2-Chloro-N-cyclohexyl-N-phenylacetamide
2-Chloro-N-cyclohexyl-N-phenylacetamide
Formel: C₁₄H₁₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD01566907
CAS-Nummer: 100721-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR