2-Formylfuran-3-boronic acid pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxaldehyde, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-furaldehyde

OR2773-1G
APOSOR2773-1GEA 253 EUR
APOSOR2773-1G
2-Formylfuran-3-boronic acid pinacol ester 98%
2-Formylfuran-3-boronic acid pinacol ester
Formel: C₁₁H₁₅BO₄
Molecular Weight: 222,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368242
CAS-Nummer: 1055881-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR