4-(1,3-Dimethylimidazolidin-2-yl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Dimethyl-2-(pyridin-4-yl)imidazolidine

OR27732-50MG
APOSOR27732-50MGEA 21.9 EUR
APOSOR27732-50MG
4-(1,3-Dimethylimidazolidin-2-yl)pyridine
4-(1,3-Dimethylimidazolidin-2-yl)pyridine
Formel: C₁₀H₁₅N₃
Molecular Weight: 177,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01570900
CAS-Nummer: 303187-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR