2,2-Dimethyl-5-nitro-2,3-dihydro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

OR27769-250MG
APOSOR27769-250MGEA 68.3 EUR
APOSOR27769-250MG
2,2-Dimethyl-5-nitro-2,3-dihydro-1H-benzimidazole
2,2-Dimethyl-5-nitro-2,3-dihydro-1H-benzimidazole
Formel: C₉H₁₁N₃O₂
Molecular Weight: 193.203 g/mol
Schmelzpunkt: 239…241 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28963764
CAS-Nummer: 306935-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR