4-(Dimethylamino)chalcone
Lieferant: Apollo Scientific
|
Achtung
|
OR27791-1G
OR27791-250MG
APOSOR27791-1GEA
0
EUR
APOSOR27791-1G
APOSOR27791-250MG
4-(Dimethylamino)chalcone
4-(Dimethylamino)chalcone
Formel:
C₁₇H₁₇NO Schmelzpunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168677 CAS-Nummer: 1030-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...