2'-Hydroxy-3,4,4'-trimethoxychalcone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[3-(3,4-Dimethoxyphenyl)acryloyl]-5-methoxyphenol, 3-(3,4-Dimethoxyphenyl)-1-(2-hydroxy-4-methoxyphenyl)prop-2-en-1-one

OR27802-250MG
APOSOR27802-250MGEA 46.4 EUR
APOSOR27802-250MG
2'-Hydroxy-3,4,4'-trimethoxychalcone
2'-Hydroxy-3,4,4'-trimethoxychalcone
Formel: C₁₈H₁₈O₅
Molecular Weight: 314,333 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00205556
CAS-Nummer: 10493-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR