1-(4-methylidenepiperidino)ethan-1-one 95+%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Methylenepiperidin-1-yl)ethanone

OR27816-250MG
APOSOR27816-250MGEA 190 EUR
APOSOR27816-250MG
1-(4-methylidenepiperidino)ethan-1-one 95+%
1-(4-methylidenepiperidino)ethan-1-one
Formel: C₈H₁₃NO
Molecular Weight: 139.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00831797
CAS-Nummer: 308087-58-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR