6-Aminouracil

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2,6-dihydroxypyrimidin, 4-Aminouracil, 6-Amino-2,4-dihydroxypyrimidin

OR27848-100G
APOSOR27848-100GEA 21.9 EUR
APOSOR27848-100G
6-Aminouracil
6-Aminouracil
Formel: C₄H₅N₃O₂
Molecular Weight: 127,10 g/mol
Schmelzpunkt: >360 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068470
CAS-Nummer: 873-83-6
EINECS: 212-854-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR