(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)ethanoic acid, 4-(Carboxymethyl)-3,4-dihydro-3-oxo-2H-1,4-benzothiazine

OR2786-1G
APOSOR2786-1GEA 161 EUR
APOSOR2786-1G
(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetic acid
(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetic acid
Formel: C₁₀H₉NO₃S
Molecular Weight: 223.248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665896
CAS-Nummer: 100637-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR