4-Amino-2,3,5-trichlorpyridin 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,5-trichloropyridin-4-amine

OR27920-250MG
APOSOR27920-250MGEA 236 EUR
APOSOR27920-250MG
4-Amino-2,3,5-trichlorpyridin 95+%
4-Amino-2,3,5-trichloropyridine
Formel: C₅H₃Cl₃N₂
Molecular Weight: 197,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153908
CAS-Nummer: 28443-69-8
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR