Pseudothiohydantoin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-aminothiazol-4(5H)-one, 2-Amino-4,5-dihydro-1,3-thiazol-4-one

OR27978-5G
APOSOR27978-5GEA 21.9 EUR
APOSOR27978-5G
Pseudothiohydantoin 97%
Pseudothiohydantoin
Formel: C₃H₄N₂OS
Molecular Weight: 116,14 g/mol
Schmelzpunkt: 249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003186
CAS-Nummer: 556-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR