(2-Brom-4-pyridinyl)methanol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR2802-25G
APOSOR2802-25GEA
133
EUR
APOSOR2802-25G
(2-Brom-4-pyridinyl)methanol 98%
(2-Brom-4-pyridinyl)methanol
Formel:
C₆H₆BrNO Schmelzpunkt: 60…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039313 CAS-Nummer: 118289-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...