1-(Fur-2-yl)-3-(4-methylphenyl)prop-2-en-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[3-(4-Methylphenyl)acryloyl]furan, 2-(4-Methylcinnamoyl)furan

OR28027-250MG
APOSOR28027-250MGEA 48 EUR
APOSOR28027-250MG
1-(Fur-2-yl)-3-(4-methylphenyl)prop-2-en-1-one
1-(Fur-2-yl)-3-(4-methylphenyl)prop-2-en-1-one
Formel: C₁₄H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00276130
CAS-Nummer: 14385-62-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR