Picloram
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-3,5,6-trichloropicolinic Acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic Acid
OR28120-1G
APOSOR28120-1GEA
0
EUR
APOSOR28120-1G
Picloram
Picloram
Formel:
C₆H₃Cl₃N₂O₂ Molecular Weight: 241,45 g/mol Siedepunkt: 421 °C Schmelzpunkt: 200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012101 CAS-Nummer: 1918-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...