4-Chloro-3-oxo-2-(quinolin-2(1H)-ylidene)butanenitrile

Lieferant: Apollo Scientific
Achtung

OR28134-100MG
APOSOR28134-100MGEA 122 EUR
APOSOR28134-100MG
4-Chloro-3-oxo-2-(quinolin-2(1H)-ylidene)butanenitrile
4-Chloro-3-oxo-2-(quinolin-2(1H)-ylidene)butanenitrile
Formel: C₁₃H₉ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00832023
CAS-Nummer: 128914-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR