6-Methylbenzo[d]oxazole 99+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methylbenzoxazole

OR28152-250MG
APOSOR28152-250MGEA 21.9 EUR
APOSOR28152-250MG
6-Methylbenzo[d]oxazole 99+%
6-Methylbenzo[d]oxazole
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 29…31 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216937
CAS-Nummer: 10531-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR