N-(Chloroacetyl)glycine ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl [(chloroacetyl)amino]acetate, Ethyl N-(chloroacetyl)glycinate

OR28155-5G
APOSOR28155-5GEA 102 EUR
APOSOR28155-5G
N-(Chloroacetyl)glycine ethyl ester
N-(Chloroacetyl)glycine ethyl ester
Formel: C₆H₁₀ClNO₃
Molecular Weight: 179,602 g/mol
Schmelzpunkt: 62…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216646
CAS-Nummer: 41602-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR