Isochinolin-4-amin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Aminoisoquinoline, Isoquinolin-4-amine
OR2818-1G
OR2818-5G
APOSOR2818-1GEA
36.4
EUR
APOSOR2818-1G
APOSOR2818-5G
Isochinolin-4-amin
Isochinolin-4-amin
Formel:
C₉H₈N₂ Schmelzpunkt: 107…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00034752 CAS-Nummer: 23687-25-4 EINECS: 245-823-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...